Ismatec® Ecoline Peristaltic Pumps
Stackable, space-saving design
  • Start, stop, and reverse flow with a single switch
  • Angled speed control displays for easy user access
  • Stainless steel housings ensure durability
These economical pumps include preassembled heads for simple setup and operation. Start, stop, and reverse flow with a single switch. Stainless steel pump housings ensure durability. Pump housings feature angled speed control displays for easy user access. Four stabilizing feet allow you to stack multiple drives. Pumps come with rotors for use with 1.6 and 2.4-mm wall tubing.
£1,219.00 - £2,004.00GBP / Each
£1,462.80 - £2,404.80GBP (incl VAT)
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Number of Channels Power (VAC)
Availability Pricing
COMPARE
5400 1 115
£1,548.00GBP / Each
£1,857.60GBP (incl VAT)
COMPARE
5400 1 230
£1,575.00GBP / Each
£1,890.00GBP (incl VAT)
COMPARE
4600 1 115
£1,553.00GBP / Each
£1,863.60GBP (incl VAT)
COMPARE
4600 1 230
£1,581.00GBP / Each
£1,897.20GBP (incl VAT)
COMPARE
5000 1 115
£1,567.00GBP / Each
£1,880.40GBP (incl VAT)
COMPARE
5000 1 230
£1,594.00GBP / Each
£1,912.80GBP (incl VAT)
COMPARE
4200 1 115
£1,575.00GBP / Each
£1,890.00GBP (incl VAT)
COMPARE
4200 1 230
£1,603.00GBP / Each
£1,923.60GBP (incl VAT)
COMPARE
1300 1 115
£1,474.00GBP / Each
£1,768.80GBP (incl VAT)
COMPARE
1300 1 230
£1,500.00GBP / Each
£1,800.00GBP (incl VAT)
COMPARE
150 8 115
£1,639.00GBP / Each
£1,966.80GBP (incl VAT)
COMPARE
150 8 230
£1,668.00GBP / Each
£2,001.60GBP (incl VAT)
COMPARE
1300 1 115
£1,595.00GBP / Each
£1,914.00GBP (incl VAT)
COMPARE
1300 1 230
£1,623.00GBP / Each
£1,947.60GBP (incl VAT)
COMPARE
1300 1 115
£1,595.00GBP / Each
£1,914.00GBP (incl VAT)
COMPARE
1300 1 230
£2,004.00GBP / Each
£2,404.80GBP (incl VAT)
COMPARE
82 4 115
£1,613.00GBP / Each
£1,935.60GBP (incl VAT)
COMPARE
150 1 115
£1,240.00GBP / Each
£1,488.00GBP (incl VAT)
COMPARE
82 4 230
£1,641.00GBP / Each
£1,969.20GBP (incl VAT)
COMPARE
150 1 230
£1,219.00GBP / Each
£1,462.80GBP (incl VAT)
Showing 1- 20 of 20