BriskHeat® Silicone Rubber Heating Tapes - 240 VAC
Moisture and chemical resistant
  • Maximum exposure temperature of 450°F (232°C)
  • Nongrounded
Solve a wide range of heating applications without risk of damage due to chemical and moisture spillover. Tapes are made of fine-strand resistance wires that are double insulated with braided fiberglass and knitted into flat tapes; then completely enclosed in high-strength, hightemperature-
resistant silicone rubber.
The 240 VAC tapes are supplied with bare leads.
Warrning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
The 240 VAC tapes are supplied with bare leads.
Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
£190.26 - £401.00GBP / Each
£228.31 - £481.20GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat 102120L Silicone Rubber Insulated Heating Tape, 12' x 1" , 624 Watts, 240V WZ-03106-29 Mfr # TBS0-102-120L
COMPARE
12 450 240
£337.10GBP / Each
£404.52GBP (incl VAT)
BriskHeat 102140L Silicone Rubber Insulated Heating Tape, 14' x 1" , 936 Watts, 240V WZ-03106-30 Mfr # TBS0-102-140L
COMPARE
14 450 240
£401.00GBP / Each
£481.20GBP (incl VAT)
BriskHeat TBSO-102-020 Silicone Rubber Insulated Heating Tape, 2' x 1" , 104 Watts, 240 V WZ-03110-21 Mfr # TBSO-102-020
COMPARE
2 500 240
£190.26GBP / Each
£228.31GBP (incl VAT)
BriskHeat TBSO-102-040 Silicone Rubber Insulated Heating Tape, 4' x 1" , 208 Watts, 240 V WZ-03110-41 Mfr # TBSO-102-040
COMPARE
4 500 240
£221.54GBP / Each
£265.85GBP (incl VAT)
BriskHeat TBSO-102-060 Silicone Rubber Insulated Heating Tape, 6' x 1" , 312 Watts, 240 V WZ-03110-61 Mfr # TBSO-102-060
COMPARE
6 500 240
£274.30GBP / Each
£329.16GBP (incl VAT)
BriskHeat BSO-052-120 Silicone Rubber Insulated Heating Tape, 12' x 1/2" , 312 Watts, 240 V WZ-03111-13 Mfr # TBSO-052-120
COMPARE
12 500 240
£256.23GBP / Each
£307.48GBP (incl VAT)
BriskHeat TBSO-052-020 Silicone Rubber Insulated Heating Tape, 2' x 1/2" , 52 Watts, 240 V WZ-03111-21 Mfr # TBSO-052-020
COMPARE
2 500 240
£268.49GBP / Each
£322.19GBP (incl VAT)
BriskHeat TBSO-052-040 Silicone Rubber Insulated Heating Tape, 4' x 1/2" , 104 Watts, 240 V WZ-03111-41 Mfr # TBSO-052-040
COMPARE
4 500 240
£244.95GBP / Each
£293.94GBP (incl VAT)
BriskHeat TBSO-052-060 Silicone Rubber Insulated Heating Tape, 6' x 1/2" , 156 Watts, 240 V WZ-03111-61 Mfr # TBSO-052-060
COMPARE
6 500 240
£248.35GBP / Each
£298.02GBP (incl VAT)
BriskHeat TBSO-052-080 Silicone Rubber Insulated Heating Tape, 8' x 1/2" , 208 Watts, 240 V WZ-03111-81 Mfr # TBSO-052-080
COMPARE
8 500 240
£219.40GBP / Each
£263.28GBP (incl VAT)
Showing 1- 10 of 10