BriskHeat® Silicone Rubber Heating Tapes - 120 VAC
Moisture and chemical resistant
  • Maximum exposure temperature of 450°F (232°C)
  • Nongrounded
Solve a wide range of heating applications without risk of damage due to chemical and moisture spillover. Tapes are made of fine-strand resistance wires that are double insulated with braided fiberglass and knitted into flat tapes; then completely enclosed in high-strength, hightemperature-
resistant silicone rubber.
The 120 VAC tapes have a same-end lead configuration and are supplied with a half plug on each end; the half plugs fit together and connect to form a standard 120 VAC two-prong plug. Silicone rubber-insulated lead wires extend two feet from opposite ends of the tape to simplify connection to power source.
Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
£155.57 - £498.41GBP / Each
£186.68 - £598.09GBP (incl VAT)
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Power (VAC) Description
Availability Pricing
BriskHeat TBSO-101-020 Silicone Rubber Insulated Heating Tape, 2' x 1" , 104 Watts, 120 V WZ-03110-20 Mfr # TBSO-101-020
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 2' x 1\" , 104 Watts, 120 V
£179.83GBP / Each
£215.80GBP (incl VAT)
BriskHeat TBSO-101-040 Silicone Rubber Insulated Heating Tape, 4' x 1" , 208 Watts, 120 V WZ-03110-40 Mfr # TBSO-101-040
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 4' x 1\" , 208 Watts, 120 V
£211.96GBP / Each
£254.35GBP (incl VAT)
BriskHeat TBSO-101-060 Silicone Rubber Insulated Heating Tape, 6' x 1" , 312 Watts, 120 V WZ-03110-60 Mfr # TBSO-101-060
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 6' x 1\" , 312 Watts, 120 V
£233.67GBP / Each
£280.40GBP (incl VAT)
BriskHeat TBSO-101-080 Silicone Rubber Insulated Heating Tape, 8' x 1" , 416 Watts, 120 V WZ-03110-80 Mfr # TBSO-101-080
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 8' x 1\" , 416 Watts, 120 V
£274.53GBP / Each
£329.44GBP (incl VAT)
BriskHeat 627 Silicone Rubber Insulated Heating Tape, 12' x 1" , Watts, 120V WZ-03110-82 Mfr # TBS0-101-120
COMPARE
450 120 Silicone Rubber Insulated Heating Tape, 12' x 1\" , Watts, 120V
£343.91GBP / Each
£412.69GBP (incl VAT)
BriskHeat 731 Silicone Rubber Insulated Heating Tape, 14' x 1" , Watts, 120V WZ-03110-84 Mfr # TBS0-101-140
COMPARE
450 120 Silicone Rubber Insulated Heating Tape, 14' x 1\" , Watts, 120V
£389.02GBP / Each
£466.82GBP (incl VAT)
BriskHeat TBSO-051-120 Silicone Rubber Insulated Heating Tape, 12' x 1/2" , 312 Watts, 120 V WZ-03111-12 Mfr # TBSO-051-120
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 12' x 1/2\" , 312 Watts, 120 V
£237.70GBP / Each
£285.24GBP (incl VAT)
BriskHeat TBSO-051-020 Silicone Rubber Insulated Heating Tape, 2' x 1/2" , 52 Watts, 120 V WZ-03111-20 Mfr # TBSO-051-020
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 2' x 1/2\" , 52 Watts, 120 V
£204.94GBP / Each
£245.93GBP (incl VAT)
BriskHeat TBSO-051-040 Silicone Rubber Insulated Heating Tape, 4' x 1/2" , 104 Watts, 120 V WZ-03111-40 Mfr # TBSO-051-040
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 4' x 1/2\" , 104 Watts, 120 V
£185.15GBP / Each
£222.18GBP (incl VAT)
BriskHeat TBSO-051-060 Silicone Rubber Insulated Heating Tape, 6' x 1/2" , 156 Watts, 120 V WZ-03111-60 Mfr # TBSO-051-060
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 6' x 1/2\" , 156 Watts, 120 V
£155.57GBP / Each
£186.68GBP (incl VAT)
BriskHeat TBSO-051-080 Silicone Rubber Insulated Heating Tape, 8' x 1/2" , 208 Watts, 120 V WZ-03111-80 Mfr # TBSO-051-080
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 8' x 1/2\" , 208 Watts, 120 V
£199.83GBP / Each
£239.80GBP (incl VAT)
BriskHeat TBSO-201-020 Silicone Rubber Insulated Heating Tape, 2' x 2" , 208 Watts, 120 V WZ-03112-20 Mfr # TBSO-201-020
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 2' x 2\" , 208 Watts, 120 V
£221.54GBP / Each
£265.85GBP (incl VAT)
BriskHeat TBSO-201-040 Silicone Rubber Insulated Heating Tape, 4' x 2" , 416 Watts, 120 V WZ-03112-40 Mfr # TBSO-201-040
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 4' x 2\" , 416 Watts, 120 V
£293.68GBP / Each
£352.42GBP (incl VAT)
BriskHeat TBSO-201-060 Silicone Rubber Insulated Heating Tape, 6' x 2" , 624 Watts, 120 V WZ-03112-60 Mfr # TBSO-201-060
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 6' x 2\" , 624 Watts, 120 V
£410.73GBP / Each
£492.88GBP (incl VAT)
BriskHeat TBSO-201-080 Silicone Rubber Insulated Heating Tape, 8' x 2" , 832 Watts, 120 V WZ-03112-80 Mfr # TBSO-201-080
COMPARE
500 120 Silicone Rubber Insulated Heating Tape, 8' x 2\" , 832 Watts, 120 V
£498.41GBP / Each
£598.09GBP (incl VAT)
Showing 1- 15 of 15