BriskHeat® Silicone-Rubber Heater Blankets Plastic Surfaces with Control and Adhesive-240 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 160°F (71°C)
  • Easy-to-apply industrial strength pressure sensitive adhesive backing
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications.
Blanket with Control offers extra convenience and functionality with its included adjustable thermostat control. Models for metal surfaces can be adjusted between 50 to 160°F . No additional control is necessary. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Models for plastic surfaces have a lower 1.25 watts/in2 power density to prevent scorching of the surface.
£213.67 - £310.07GBP / Each
£256.40 - £372.08GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRP06242PADJB Silicone Heating Blanket with Controller, 50-160F, 6x24 Size, 240 Volt, 180 Watt WZ-36002-55 Mfr # SRP06242PADJB
COMPARE
24 160 240
£213.67GBP / Each
£256.40GBP (incl VAT)
BriskHeat SRP12242PADJB Silicone Heating Blanket with Controller, 50-160F, 12x24 Size, 240 Volt, 360 Watt WZ-36002-61 Mfr # SRP12242PADJB
COMPARE
24 160 240
£310.07GBP / Each
£372.08GBP (incl VAT)