BriskHeat® Silicone-Rubber Heater Blankets Plastic Surfaces with Control-240 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 160°F (71°C)
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications.
Blanket with Control offers extra convenience and functionality with its included adjustable thermostat control. Models for metal surfaces can be adjusted between 50 to 160°F . No additional control is necessary. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Models for plastic surfaces have a lower 1.25 watts/in2 power density to prevent scorching of the surface.
£155.57 - £471.59GBP / Each
£186.68 - £565.91GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRP06122ADJB Silicone Heating Blanket with Controller, 90 W, for Plastics, 6" x 12" , 240 V WZ-36002-73 Mfr # SRP06122ADJB
COMPARE
12 160 240
£155.57GBP / Each
£186.68GBP (incl VAT)
BriskHeat SRP06362ADJB Silicone Heating Blanket with Controller, 270 W, for Plastics, 6" x 36" , 240 V WZ-36002-77 Mfr # SRP06362ADJB
COMPARE
36 160 240
£256.23GBP / Each
£307.48GBP (incl VAT)
BriskHeat SRP12242ADJB Silicone Heating Blanket with Controller, 360 W, for Plastics, 12" x 24" , 240 V WZ-36002-81 Mfr # SRP12242ADJB
COMPARE
24 160 240
£280.49GBP / Each
£336.59GBP (incl VAT)
BriskHeat SRP12362ADJB Silicone Heating Blanket with Controller, 540 W, Plastic Surface, 12" x 36" , 240 V WZ-36002-83 Mfr # SRP12362ADJB
COMPARE
36 160 240
£417.75GBP / Each
£501.30GBP (incl VAT)
BriskHeat SRP24242ADJB Silicone Heating Blanket with Controller, 720 W, for Plastics, 24" x 24" , 240 V WZ-36002-89 Mfr # SRP24242ADJB
COMPARE
24 160 240
£471.59GBP / Each
£565.91GBP (incl VAT)