BriskHeat® Silicone-Rubber Heater Blankets Plastic Surfaces - 240 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 450°F (232°C)
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 watts /in2 power density.
£125.14GBP / Each
£150.17GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRP12122 Silicone Heating Blanket, 12x12 Size, 240 Volt, 180 Watt, for plastic surfaces WZ-36001-85 Mfr # SRP12122
COMPARE
12 450 240
£125.14GBP / Each
£150.17GBP (incl VAT)