Shimpo High Capacity Digital Force Gauges
No more reading upside down or craning your neck
  • Reversible display allows use as a handheld and mounted upside down
  • Dual-labeled keypad ensures easy operation in any orientation
  • Rechargeable batteries and AC line voltage compatibility increase versatility
  • View current force measurement or peak force for the test
These high-capacity force gauges feature rugged construction and simple one-button operation while offering accurate results. The fast sampling rate provides enhanced test resolution, ideal for quality inspections, finished goods testing, or research and development. The 180° reversible display with dual-printed keypad makes use as either a handheld or a mounted gauge convenient and easy.
Choose the FGV series if you need to record and export test data to PC or if you require a built-in Hi-Lo comparator function to provide Go/No-Go testing. These models feature onboard memory for 1000 data points and offer the flexibility of using three outputs: USB, RS-232, and analog. Software is available for free download from the manufacturer's website.
£868.02 - £1,016.63GBP / Each
£1,041.62 - £1,219.96GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Output
Availability Pricing
Shimpo FGE-500HXY High Capacity Digital Force Gauge, 500 lb WZ-25301-13 Mfr # FGE-500HXY
COMPARE
500 None
£868.02GBP / Each
£1,041.62GBP (incl VAT)
Shimpo FGV-500HXY High Capacity Digital Force Gauge with Output, 500 lb WZ-25302-91 Mfr # FGV-500HXY
COMPARE
500 USB, RS-232, and analog
£1,016.63GBP / Each
£1,219.96GBP (incl VAT)