This product is now discontinued, View Related Products
Trajan SGE eVol® Digital Analytical Syringe Starter Kits
World's first digital analytic syringes combine precision and speed
  • Program and store up to 98 steps of workflow
  • Password protection options for improved security
  • Save calibrations for each syringe, enabling standards compliance
  • Suitable for direct injection onto a chromatography column with consistent flow rate
  • Be safer when working with volatile samples — aliquots are dispensed into sealed vials
eVol XR syringes combine a digitally controlled electronic drive and XCHANGE® enabled analytics for improved accuracy and reproducibility. The eVol XR system allows quick and simple syringe changes, so syringes can be dedicated to types of samples or methods, helping prevent reagent cross-contamination. And calibration factors can be easily programmed and stored for each syringe, enabling compliance with global lab standards. Syringe programming can be password protected for security.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Needle Length (in) Needle Tip Style Description
Availability Pricing
Trajan SGE eVoL XR Electronic Syringe Starter Kit WZ-17940-22 Mfr # 2910200
COMPARE
1.969 Bevel EVOL XR Electronic Syringe Starter Kit
Call For Price
Trajan SGE eVoL Electronic Syringe Starter Kit; NMR Edition WZ-17940-24 Mfr # 2910100
COMPARE
1.969 Bevel EVOL Electronic Syringe Starter Kit; NMR Edition