This product is now discontinued, View Related Products
Servus® by Honeywell™ 12”H Polyurethane Processing Overboots
Protect yourself against abrasions, slipping, and splashes
  • Completely waterproof Chemical-resistant polyurethane construction Light weight prevents foot and leg fatigue Easily put on and take off Unique strap system provides a secure fit to stop boot from slipping Available in mens’ sizes XS to 3XL
Servus® by Honeywell’s™ processing overboots are constructed of heavy-duty 0.8 mm thermal polyurethane to keep your feet dry. The 100% waterproof design offers splash protection against water and most chemicals for longer wear. Easily put on and take off using the extra-large gusset. The one-of-a-kind open tread design allows for firm footing in wet and oily conditions. The ankle strap provides a snug, adjustable fit for your needs, and its snap closure allows for easy sanitation. Blue and white color; with soft toe.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Color Height (in) Men's Size
Availability Pricing
Honeywell Servus Polyurethane Processing Overboots, XS; 1 Pair WZ-86326-71 Mfr # IT12PT17-BLU-XSM
COMPARE
White & blue 12 XS
Call For Price
Honeywell Servus Polyurethane Processing Overboots, S; 1 Pair WZ-86326-72 Mfr # IT12PT17-BLU-SML
COMPARE
White & blue 12 S
Call For Price
Honeywell Servus Polyurethane Processing Overboots, M; 1 Pair WZ-86326-73 Mfr # IT12PT17-BLU-MED
COMPARE
White & blue 12 M
Call For Price
Honeywell Servus Polyurethane Processing Overboots, L; 1 Pair WZ-86326-74 Mfr # IT12PT17-BLU-LRG
COMPARE
White & blue 12 L
Call For Price
Honeywell Servus Polyurethane Processing Overboots, XL; 1 Pair WZ-86326-75 Mfr # IT12PT17-BLU-1XL
COMPARE
White & blue 12 XL
Call For Price
Honeywell Servus Polyurethane Processing Overboots, 2XL; 1 Pair WZ-86326-76 Mfr # IT12PT17-BLU-2XL
COMPARE
White & blue 12 2XL
Call For Price
Honeywell Servus Polyurethane Processing Overboots, 3XL; 1 Pair WZ-86326-77 Mfr # IT12PT17-BLU-3XL
COMPARE
White & blue 12 3XL
Call For Price
Showing 1- 7 of 7