Self-Closing Threaded Diaphragm Check Valves, 1/2" NPT(F)
Patented diaphragm design does not depend on gravity, mounting position, or reverse flow to seal
  • Leak-free sealing prevents reverse flow
  • Create a secure seal using pipe thread connections
  • Diaphragm automatically positions itself against seat in the identical location
  • Diaphragm begins to open at approximately 1 to 1.5 psi
  • Flushing action of the inlet flow keeps the sealing area clean
  • 1/2" NPT(F) x 1/2" NPT(F) connections
Use the self-closing diaphragm check valves to protect against hazardous and corrosive reverse flow. The diaphragm reseats against the seal without relying on gravity, mounting position, or reverse flow, ensuring reliable sealing even at low pressures. A union nut allows easy inspection and removal with minimal piping disruption. No loose or sliding internal parts eliminate chattering and help keep operation quiet. As inlet flow passes through the valve, both the diaphragm and valve seat are kept clean by the flushing action, keeping the sealing area clean and free of particles, minimizing the risk of leakage.
£103.80 - £282.60GBP / Each
£124.56 - £339.12GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Housing Material Inlet Port Outlet Port
Availability Pricing
Self-Closing Check Valve, PVDF, 1/2" NPT(F) 4852714 Mfr # CKM050V-PF
COMPARE
PVDF 1/2" NPT(F) 1/2" NPT(F)
£282.60GBP / Each
£339.12GBP (incl VAT)
Self-Closing Check Valve, PP, 1/2" NPT(F) 4852715 Mfr # CKM050V-PP
COMPARE
Polypropylene 1/2" NPT(F) 1/2" NPT(F)
£149.25GBP / Each
£179.10GBP (incl VAT)
Self-Closing Check Valve, PVC, 1/2" NPT(F) 4852716 Mfr # CKM050V-PV
COMPARE
PVC 1/2" NPT(F) 1/2" NPT(F)
£103.80GBP / Each
£124.56GBP (incl VAT)