Scienceware Spinbar Octagon Rare Earth Stirbars
Samarium Cobalt magnet contains PTFE which provides strong coupling with drive magnets to reduce frequency of spinout. Ideal for viscous solutions, high speed stirring and extended stirring applications. Octagon shape with pivot ring creates strong turbulence with minimal clicking and friction.
£12.58 - £40.37GBP / Each
£15.10 - £48.44GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in)
Availability Pricing
Scienceware F371020003 Spinbar Rare Earth Oct 3" X 1/2" WZ-04775-00 Mfr # F371020003
COMPARE
0.5 3
£40.37GBP / Each
£48.44GBP (incl VAT)
Scienceware F371020138 Spinbar Rare Earth Oct 1" X 3/8" WZ-04775-02 Mfr # F371020138
COMPARE
0.375 1
£17.30GBP / Each
£20.76GBP (incl VAT)
Scienceware F371020238 Spinbar Rare Earth Oct 2" X 3/8" WZ-04775-04 Mfr # F371020238
COMPARE
0.375 2
£35.12GBP / Each
£42.14GBP (incl VAT)
Scienceware F371021238 Spinbar Rare Earth Oct 1/2 X 3/8 WZ-04775-06 Mfr # F371021238
COMPARE
0.375 1/2
£12.58GBP / Each
£15.10GBP (incl VAT)