Scienceware SATURN SPINBAR
Coated in PTFE, stir bar contains a large sphere in center of stir bar that lifts the stirring bar ends while spinning and reducing the surface contact area to allow stir bar to spin with decreased drag. AUTOCLAVABLE at 121°C (250°F)
£30.39 - £43.84GBP / Each
£36.47 - £52.61GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Scienceware F371160002 Saturn Spinbar, Magnetic Stir Bar, 40 mm L; 1/Pk WZ-04775-24 Mfr # F371160002
COMPARE
Saturn Spinbar, Magnetic Stir Bar, 40 mm L; 1/Pk
£30.39GBP / Each
£36.47GBP (incl VAT)
Scienceware F371160003 Saturn Spinbar, Magnetic Stir Bar, 50 mm L; 1/Pk WZ-04775-26 Mfr # F371160003
COMPARE
Saturn Spinbar, Magnetic Stir Bar, 50 mm L; 1/Pk
£43.84GBP / Each
£52.61GBP (incl VAT)