Sartorius Secura Toploading Balances
Designed for users who work in regulated environments and require high accuracy
  • Color touch screen with picture graphic menu
  • Optoelectronic level sensor with alarm function and user interface
  • isoCAL offers fully automatic, motorized calibration and adjustment
  • No weighing below the minimum value with programmable SQmin
  • Easy to clean with removable glass draft shield, stain-resistant surfaces, and smooth contours
  • Supervisor lock prevents unintentional menu changes
  • Mini USB interface port enables communication with a PC or printer
  • Weighing applications include general weighing, density, percentage, check weighing, peak hold, counting, and averaging
  • Built-in overload protection
£2,193.03 - £3,814.81GBP / Each
£2,631.64 - £4,577.77GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Sartorius SECURA513-1S Toploading Balance, 510g x 0.001g, Internal Calibration WZ-11800-67 Mfr # SECURA513-1S
COMPARE
510 100 to 240 0.001
£3,563.70GBP / Each
£4,276.44GBP (incl VAT)
Sartorius SECURA5102-1S Toploading Balance, 5100g x 0.01g, Internal Calibration WZ-11800-71 Mfr # SECURA5102-1S
COMPARE
5100 100 to 240 0.01
£3,598.81GBP / Each
£4,318.57GBP (incl VAT)
Sartorius SECURA3102-1S Toploading Balance, 3100g x 0.01g, Internal Calibration WZ-11800-72 Mfr # SECURA3102-1S
COMPARE
3200 100 to 240 0.01
£3,130.22GBP / Each
£3,756.26GBP (incl VAT)
Sartorius SECURA2102-1S Toploading Balance, 2100g x 0.01g, Internal Calibration WZ-11800-73 Mfr # SECURA2102-1S
COMPARE
2200 100 to 240 0.01
£2,886.55GBP / Each
£3,463.86GBP (incl VAT)
Sartorius Secura613-1S Toploading Balance 610g x 0.001g, Internal Calibration WZ-11955-01 Mfr # SECURA613-1S
COMPARE
610 115 0.001
£3,702.35GBP / Each
£4,442.82GBP (incl VAT)
Sartorius Secura612-1S Toploading Balance 610g x 0.01g, Internal Calibration WZ-11955-02 Mfr # SECURA612-1S
COMPARE
610 115 0.01
£2,193.03GBP / Each
£2,631.64GBP (incl VAT)
Sartorius Secura6102-1S Toploading Balance 6100g x 0.01g, Internal Calibration WZ-11955-03 Mfr # SECURA6102-1S
COMPARE
6100 115 0.01
£3,814.81GBP / Each
£4,577.77GBP (incl VAT)
Showing 1- 7 of 7