This product is now discontinued, View Related Products

Sartorius Entris Analytical Balances

Extraordinary accuracy combined with everyday features for a competitive price
  • Perform basic weighing with below balance, counting, percent, animal weighing, and density determination functions
  • Anti-theft feature secures balance to work station
  • Functional in 7 languages
  • Document critical weights via RS-232 communication with PC or printer
  • Sliding doors and removable shield chamber parts make cleanup easy
  • Backlit high-contrast display delivers excellent visibility in any lighting condition
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Sartorius ENTRIS64I-1SUS Analytical Balance 60 x 0.1mg, Internal Calibration WZ-11952-19 Mfr # ENTRIS64I-1SUS
COMPARE
Internal 60 0.0001
Call For Price
Sartorius ENTRIS124I-1SUS Analytical Balance 120 g x 0.1mg, Internal Calibration WZ-11952-21 Mfr # ENTRIS124I-1SUS
COMPARE
Internal 120 0.0001
Call For Price
Sartorius ENTRIS224I-1SUS Analytical Balance 220 g x 0.1mg, Internal Calibration WZ-11952-22 Mfr # ENTRIS224I-1SUS
COMPARE
Internal 220 0.0001
Call For Price
Sartorius ENTRIS64-1SUS Analytical Balance 60 x 0.1mg, External Calibration WZ-11955-19 Mfr # ENTRIS64-1SUS
COMPARE
External 60 0.0001
Call For Price
Sartorius ENTRIS124-1SUS Analytical Balance 120g x 0.1mg, External Calibration WZ-11955-21 Mfr # ENTRIS124-1SUS
COMPARE
External 120 0.0001
Call For Price
Sartorius ENTRIS224-1SUS Analytical Balance 220g x 0.1mg, External Calibration WZ-11955-22 Mfr # ENTRIS224-1SUS
COMPARE
External 220 0.0001
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat