Sartorius® Cubis® II (MCA) Analytical Balance with Software
Long-term flexibility for regulatory and laboratory needs
  • Fully customizable hardware, software, and connectivity
  • Complete data integrity for confident compliance
  • Error-free operation by assessing and correcting relevant factors
  • Advanced MCA user interface with swipe functionality
  • All-inclusive software package included
Sartorius® Cubis® II MCA is a completely customizable balance to align with your unique application and compliance requirements while ensuring reliable results. Innovative technology features draft shield technology, fast and easy motorized leveling, an integrated ionizer* for automatic electrostatic elimination, and climate sensor for environmental monitoring to ensure error-free operation.
Large 7” color touch screen display allows you to touch, scroll, and swipe with factory-installed weighing applications. The status center shows all information about your balance and environmental conditions as well as detailed help and support in case of warnings and errors.
The all-inclusive software, QApp 99, includes four different sub-packages for compliance, complex weighing applications, weighing applications and helpful tools, and connectors for data exchange. Full end-to-end data integrity follows ALCOA+ principles. Data cannot be manipulated with the user management function and LIMS connectivity. User management includes centralized password management support, high security management configuration, and login rules. Electronic signatures are in compliance with 21 CFR Part 11.
*Select models
£10,112.73 - £17,994.05GBP / Each
£12,135.28 - £21,592.86GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Sartorius MCA124SU-S00 Cubis® II Analytical Balance, 120 g x 0.1 mg; Manual Glass Draft Shield WZ-39742-49 Mfr # MCA124SU-S00
COMPARE
120 0.1 Cubis II Analytical Balance 120g X 0.1 mg with Pre-configured QApp 99 Software and Manual Glass Draft Shield
£10,112.73GBP / Each
£12,135.28GBP (incl VAT)
Sartorius MCA524SI-S00 Cubis® II Analytical Balance, 520 g x 0.1 mg; Auto Glass Draft Shield & Integrated Ionizer WZ-39742-50 Mfr # MCA524SI-S00
COMPARE
520 0.1 Cubis II Analytical Balance 520g X 0.1 mg with Pre-configured QApp 99 Software, Auto. Glass Draft shield & Integrated Ionizer
£17,994.05GBP / Each
£21,592.86GBP (incl VAT)