Sanitary Bottles, Stainless Steel
Keep air and liquids out
Sanitary bottles are made from 316L stainless steel with a 10 Ra finish standard on interior. Bottles feature a sanitary Tri-Clamp® flange opening; include an endcap, platinum-cured silicone gasket, and heavy-duty clamp.
£1,185.38 - £2,283.10GBP / Each
£1,422.46 - £2,739.72GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Eagle Stainless Stainless Steel Sanitary Container; 1L, 2" Flange WZ-07506-01 Mfr # PS-10F
COMPARE
1 2\" Stainless Steel
£1,185.38GBP / Each
£1,422.46GBP (incl VAT)
Eagle Stainless Stainless Steel Sanitary Container; 2L, 2" Flange WZ-07506-02 Mfr # PS-12F
COMPARE
2 2\" Stainless Steel
£1,442.24GBP / Each
£1,730.69GBP (incl VAT)
Eagle Stainless Stainless Steel Sanitary Container; 5L, 2" Flange WZ-07506-05 Mfr # PS-18F
COMPARE
5 2\" Stainless Steel
£1,847.48GBP / Each
£2,216.98GBP (incl VAT)
Eagle Stainless Stainless Steel Sanitary Container; 1L, 3" Flange WZ-07506-31 Mfr # BTB-10
COMPARE
1 3\" Stainless Steel
£1,280.83GBP / Each
£1,537.00GBP (incl VAT)
Eagle Stainless Stainless Steel Sanitary Container; 2L, 4" Flange WZ-07506-32 Mfr # BTB-12
COMPARE
2 4\" Stainless Steel
£1,546.37GBP / Each
£1,855.64GBP (incl VAT)
Eagle Stainless Stainless Steel Sanitary Container; 5L, 4" Flange WZ-07506-35 Mfr # BTB-16
COMPARE
5 4\" Stainless Steel
£1,767.65GBP / Each
£2,121.18GBP (incl VAT)
Eagle Stainless Stainless Steel Sanitary Container; 10L, 4" Flange WZ-07506-80 Mfr # BTB-21
COMPARE
10 4\" Stainless Steel
£1,858.77GBP / Each
£2,230.52GBP (incl VAT)
Eagle Stainless Stainless Steel Sanitary Container; 20L, 4" Flange WZ-07506-90 Mfr # BTB-27
COMPARE
20 4\" Stainless Steel
£2,283.10GBP / Each
£2,739.72GBP (incl VAT)
Showing 1- 8 of 8