Salisbury by Honeywell Insulated Tool Rolls and Sets
Helps workers meet OSHA requirements
  • Rated for exposure up to 1000 VAC and dielectrically tested at 10,000 VAC
  • Meet or exceed ASTM 1505-07 and IEC 900 Standards for Insulated Hand Tools
  • Helps comply with OSHA 1910.333 (c)(2), and NFPA 70E 2004
These quality insulated tools are made with impact-resistant and flame-resistant material. Two-color insulation makes inspection easier which adds to the overall safety. If the yellow under layer is showing, the tool may no longer be insluated properly and should be considered for replacement and shall be removed from service.
Custom kits and individual tools available. Call our Application Specialists for more information.
£566.37GBP / Each
£679.64GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Number of Pieces per Set Set Includes Description
Availability Pricing
COMPARE
9 3 slotted screwdrivers: 3⁄16 x 4'', 3⁄16 x 6'', 1⁄4 x 6'', 2 Phillips® screwdrivers: #1 x 3'', #2 x 4'', 7'' needle nose plier, 7 1⁄2'' diagonal cutting plier, 9'' Linesman’s plier 9 Piece Insulated Basic Electrician Roll
£566.37GBP / Each
£679.64GBP (incl VAT)