Rubbermaid Low-Cost Service Carts

Carts are steam cleanable
  • Premium ergonomic handle for improved control
  • Area for tools and cord management
Carts come with two seamless shelves with supports, smooth corners, molded handle, and 5" dia. rubber caster (two swivel). Each shelf measures 2-3/4" deep to contain spills. Convert to three-shelf carts with optional middle shelves, sold separately.
£390.14 - £442.30GBP / Each
£468.17 - £530.76GBP (incl VAT)

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Shelves Description
Availability Pricing
Rubbermaid 4500-88 BLACK Low-Cost Plastic Service Cart; 30" x 16" x 32-1/4" 0934800 Mfr # 4500-88 BLACK
COMPARE
2 4500-88 Low-cost plastic service cart; 30" x 16" x 32-1/4"
£442.30GBP / Each
£530.76GBP (incl VAT)
Rubbermaid 4520-88 BLACK 4520-88 Low-cost plastic service cart; 36" x 24" x 32-1/4" 0934810 Mfr # 4520-88 BLACK
COMPARE
2 4520-88 Low-cost plastic service cart; 36" x 24" x 32-1/4"
£390.14GBP / Each
£468.17GBP (incl VAT)
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat