Rex Gauge Surface Testing Accessories
Optimize performance of your Rex Gauge durometer
  • Test stands improve ease of use
Test stands provide a convenient and accurate way to perform repeated hardness tests. Use stands to test pressure uniformly for compliance with ASTM D-2240 and eliminate human error.
The AC adapter is suitable for use with 220 VAC power input.
£1,802.98GBP / Each
£2,163.58GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Rex Gauge OS-3 Accessory Test stand for micro A durometers WZ-59792-64 Mfr # OS-3
COMPARE
Accessory Test stand for micro A durometers
£1,802.98GBP / Each
£2,163.58GBP (incl VAT)