Cole-Parmer® Essentials Reversible Microcentrifuge Tube Racks, PP
Withstand temperatures down to -80 °C (-112 °F)
  • Autoclavable
One side of this polypropylene (PP) racks is designed to hold 0.5mL microcentrifuge tubes; the other side holds 1.5 to 2.0mL tubes. Withstand temperatures down to -80°C (-112°F).
£17.97 - £37.23GBP / Pkg of 5
£21.56 - £44.68GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Description
Availability Pricing
COMPARE
Assorted Reversible 96-well microcentrifuge tube rack, assorted colors
£37.23GBP / Pkg of 5
£44.68GBP (incl VAT)
COMPARE
Translucent Cover for 96-well microcentrifuge tube racks
£17.97GBP / Pkg of 5
£21.56GBP (incl VAT)