Idex® Stainless Steel Bottom-of-the-Bottle™ Solvent Filters
Protect your system from damaging particulates
  • Economical—stainless steel filter cups are replaceable
Stainless steel bottom-of-the-bottle filters allow solvent to be drawn from within 1/8” of the bottom of your solvent bottle. Filters are constructed of PEEK, ETFE, and SS.
£92.48 - £101.48GBP / Each
£110.98 - £121.78GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connections Pore Size (µm)
Availability Pricing
Idex Bottom-of-the-Bottle™ Inlet Solvent Filter Assembly, SS/PEEK/ETFE, 2 µm, 1/8" OD Tubing; 1/EA WZ-42700-00 Mfr # A-551
COMPARE
1/8\" OD tubing
£101.48GBP / Each
£121.78GBP (incl VAT)
Idex Bottom-of-the-Bottle™ Inlet Solvent Filter Assembly, SS/PEEK/ETFE, 10 µm, 1/8" OD Tubing; 1/EA WZ-42700-02 Mfr # A-550
COMPARE
1/8\" OD tubing
£92.48GBP / Each
£110.98GBP (incl VAT)