This product is now discontinued, View Related Products
Red Lion General Industrial Service Encoders
High pulse per revolution for accuracy
  • Current sink outputs
  • High pulse per revolution (PPR) rates
  • Up to 1200 PPR for fine, high-resolution counting or precision speed measurement from slow shaft speeds
  • Quadrature output for position measurement, bi-directional counting and systems with backlash counting requirements.
The units contain an L.E.D. light source and a photo sensor that scans a shaft mounted, slotted disc. An internal pulse-shaping amplifier circuit delivers a rectangular pulse signal from the current sinking output in response to the passing slots as it rotates. They can be direct-coupled to a machine shaft by means of a flexible-bellows, spring, or rubber sleeve type coupling that allows for axial and radial misalignment. They can also be coupled with light instrument timing-belts. Timing-belt drives also allow convenient gear-up or gear-down speed ratio changes that can be useful for obtaining non-standard PPR rates.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Actuator Type Max Operating Temperature (° C) Max Supply Voltage (VDC)
Availability Pricing
Red Lion ZBG01002 Single Channel General Service 6-Pin MS Connector 100PPR WZ-68493-44 Mfr # ZBG01002
COMPARE
100 PPR 100 28
Call For Price
Red Lion ZBG10002 Single Channel General Service 6-Pin MS Connector 1000PPR WZ-68493-46 Mfr # ZBG10002
COMPARE
1000 PPR 100 28
Call For Price
Red Lion ZBH01002 Dual Channel General Service 6-Pin MS Connector 100PPR WZ-68493-48 Mfr # ZBH01002
COMPARE
100 PPR 100 28
Call For Price
Red Lion ZBH10002 Dual Channel General Service 6-Pin MS Connector 1000PPR WZ-68493-50 Mfr # ZBH10002
COMPARE
1000 PPR 100 28