This product is now discontinued, View Related Products
Red Fiberglass Trays
Differentiate your samples with these red trays
Rugged fiberglass trays withstand temperature from 0 to 180°F (-17 to 83°C) under normal use. Autoclavable to 500° F (260°C) for short intervals.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Overall Height (in) Overall Length (in)
Availability Pricing
COMPARE
3/4 13
Call For Price
COMPARE
3/4 16
Call For Price
COMPARE
3/4 15
Call For Price
COMPARE
3/4 18