This product is now discontinued, View Related Products
Raven Wine Sediment Interface Detector
Predict settling and rack the must at the optimum time
  • Improve wine quality and consistency — fine tune and validate your process
  • Verify adjustments to reduce off-odors, yeast viability, and acetic acid production
  • Move the process from crush to fermentation without a hitch—save on berry and labor costs!
  • Eliminate thick sediment layers and reduce filtration needed for reclaiming
  • USA FDA compliant probe and cable materials
Determine the optimum time to rack your must! Racking too soon leads to increased risk of excess suspended solids, which causes excess fusel alcohol production, impaired fruitiness, and hydrogen sulfide/off-odor production. Rack too late or remove too many suspended solids, and end up causing fermentation in the clarification tank, slowing down fermentation, and reducing yeast viability.
The infrared sensor probe detects changes in density and sends the reading to the portable handheld meter. The probe has an integrated cable in either 20- or 50- ft lengths with bright-yellow depth markings for high visibility. Additionally, the stainless steel probe housing and polyurethane-coated cable are rugged, easy to clean, and made of USA FDA compliant materials.
The battery-operated portable meter has an IP67 weatherproof housing and easily accepts replacement probes. The ergonomic design fits in one hand or hangs on the fermentation tank. The integrated reel also helps to keep the probe cable tidy when not in use or taking shallow measurements.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description Length (in) Power
Availability Pricing
Raven Environmental SID-25200-20 Wine Sediment Interface Detector with 20 Foot Cable WZ-05956-60 Mfr # SID-20200-20
COMPARE
Wine Sediment Interface Detector with 20 Foot Cable 240 6x AA batteries (included)
Call For Price
Raven Environmental SID-25200-50 Wine Sediment Interface Detector with 50 Foot Cable WZ-05956-64 Mfr # SID-25200-50
COMPARE
Wine Sediment Interface Detector with 50 Foot Cable 600 6x AA batteries (included)