This product is now discontinued, View Related Products
Ralston Benchtop Pressure Calibration Manifolds and fittings
Save valuable time by testing multiple instruments at once
  • Calibrate multiple pressure gauges, transmitters, or transducers at once—up to 5000 psi (350 bar)
  • Vertically mounted ports spaced to accommodate larger gauges
  • Ports compatible with all Ralston Quick-test NPT, BSP, and flare gauge adapters for fast and easy tool-free changes
  • Excellent for calibrating multiple pressure instruments in a thermal chamber
Use the Ralston Quick-test™ benchtop manifolds to test multiple instruments simultaneously, saving you time and money! The manifolds are constructed of powder-coated steel for durability and strength. The stainless steel Quick-test fittings allow instruments under test to be connected easily and quickly without tools. A second side connection can be used to daisy-chain multiple manifolds together.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Min Pressure Description
Availability Pricing
Ralston Instruments MF2S-QM-QM-PL Ralston 2-Port Manifold w/Quick-Test Connections, 5000 psi WZ-16107-07 Mfr # MF2S-QM-QM-PL
COMPARE
5000 psi (345 bar) 0 psi (0 bar) Ralston 2-Port Manifold w/Quick-Test Connections, 5000 psi
Call For Price
Ralston Instruments MF4S-QM-QM-PL Ralston 4-Port Manifold w/Quick-Test Connections, 5000 psi WZ-16107-08 Mfr # MF4S-QM-QM-PL
COMPARE
5000 psi (345 bar) 0 psi (0 bar) Ralston 4-Port Manifold w/Quick-Test Connections, 5000 psi