This product is now discontinued, View Related Products

Rack Support System - Frame Clamps

Glass fiber-reinforced nylon and stainless steel components provide enormous strength
  • This material has a melting point of 221°C.
  • Resistance to salt solutions and solvents.
Use with rods, equipment clamps, or adapters to position your equipment exactly as you want. Thumbscrews are included.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Accepts Rod Diameter (in) Description
Availability Pricing
Harvard Apparatus 53-2022W Frame Clamps, Nylon and SS, open-sided, 5/Pk WZ-08024-28 Mfr # 53-2022W(X5 PIECES)
COMPARE
0.5 Frame Clamps, Nylon and Ss, Open-Sided, 5/Pk
Call For Price
Harvard Apparatus 53-2032W Frame Clamps, nylon and SS, t-joint, 5/pk WZ-08024-29 Mfr # 53-2032W(X5 PIECES)
COMPARE
0.5 Frame Clamps, nylon and SS, t-joint, 5/pk
Call For Price
Harvard Apparatus 53-2042W Frame Clamps, Nylon and SS in-Line, 5/Pk WZ-08024-30 Mfr # 53-2042W(X5 PIECES)
COMPARE
0.5 Frame Clamps, Nylon and Ssin-Line, 5/Pk
Call For Price
Harvard Apparatus 53-2012W Frame Clamps, nylon and SS, parallel, 5/pk WZ-08024-31 Mfr # 53-2012W(X5 PIECES)
COMPARE
0.5 Frame Clamps, nylon and SS, parallel, 5/pk
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat