QRP Latex Antistatic Finger Cots
Minimize charge generation
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
Latex finger cots are washed, powder-free, and treated with an anti-static agent. Use to handle Class II static-sensitive devices with thresholds above 1000 V. Average surface resistivity is 5 X 1012 ohms. Each pack is double-bagged in ESD-safe packaging to prevent charge generation.
Compliance: ISO Class 5
£29.24GBP / Pkg of 720
£35.09GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Size
Availability Pricing
QRP 7C SMALL Latex Antistatic Finger Cots, small WZ-86283-00 Mfr # 7C SMALL
COMPARE
Small
£29.24GBP / Pkg of 720
£35.09GBP (incl VAT)
QRP 7C MEDIUM Latex Antistatic Finger Cots, medium WZ-86283-01 Mfr # 7C MEDIUM
COMPARE
Medium
£29.24GBP / Pkg of 720
£35.09GBP (incl VAT)
QRP 7C LARGE Latex Antistatic Finger Cots, large WZ-86283-02 Mfr # 7C LARGE
COMPARE
Large
£29.24GBP / Pkg of 720
£35.09GBP (incl VAT)
QRP 7C XLARGE Latex Antistatic Finger Cots, extra-large WZ-86283-03 Mfr # 7C XLARGE
COMPARE
X-Large
£29.24GBP / Pkg of 720
£35.09GBP (incl VAT)