QLA Vessels Compatible with Agilent, Distek, and Hanson Dissolution Apparatus
QLA vessels provide more precise and consistent dissolution results
  • Unique serial number is laser marked on plastic rim of each vessel
  • Plastic rims help minimize glass breakage
  • 1000 mL capacity
Glass vessels offer good thermal conductivity and are highly resistance to scratching. Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle.
£232.57 - £376.92GBP / Each
£279.08 - £452.30GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Bottle Color Material Special Features
Availability Pricing
QLA Standard Vessel with Plastic Rim, 1000 mL, Amber Glass; Serialized WZ-41306-28 Mfr # GLA900-AUNP
COMPARE
Amber Glass Standard with plastic rim
£278.02GBP / Each
£333.62GBP (incl VAT)
QLA Standard Vessel with Plastic Rim, 1000 mL, Clear Glass; Serialized WZ-41306-40 Mfr # GLA900-UNP
COMPARE
Clear Glass Standard with plastic rim
£232.57GBP / Each
£279.08GBP (incl VAT)
QLA PEAK Vessel with Plastic Rim, 1000 mL, Amber Glass; Serialized WZ-41306-51 Mfr # GLA9PK-AUNP
COMPARE
Amber Glass PEAK style with plastic rim
£376.92GBP / Each
£452.30GBP (incl VAT)
QLA PEAK Vessel with Plastic Rim, 1000 mL, Clear Glass; Serialized WZ-41306-55 Mfr # GLA9PK-UNP
COMPARE
Clear Glass PEAK style with plastic rim
£299.40GBP / Each
£359.28GBP (incl VAT)