Pyrex Brand 2122 Economy Grade Burettes (Burets)
Features durable colored markings
  • Maintain fluid integrity within burette (buret) with dust cover
Economical burettes (burets) are supplied with 2mm straight bore PTFE stopcock plugs. Ideal when accuracy of the Class A model is not critical. Microfinish of the barrel provides a precision fit. Tips are carefully drawn for accuracy to insure proper drainage rate.
£188.56 - £196.99GBP / Each
£226.27 - £236.39GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Tolerance
Availability Pricing
Pyrex 2122A-50 2122 Class B Glass Burette, 50 mL; 1/Pk WZ-34544-54 Mfr # 2122A-50
COMPARE
50 0.1
£188.56GBP / Each
£226.27GBP (incl VAT)
Pyrex 2122A-100 2122 Class B Glass Burette, 100 mL; 1/Pk WZ-34544-56 Mfr # 2122A-100
COMPARE
100 0.2
£196.99GBP / Each
£236.39GBP (incl VAT)