Pyrex Brand 2094 Large Volume Burettes (Burets)
Handle more fluid
  • Maintain fluid integrity within burette (buret) with dust cap
Burettes (burets) are ideal for those requiring a larger capacity — up 1000 mL. These Class B dispensing burettes are equiped with a PTFE 2-mm bore stopcock plug to reduce freezing and lubricant contamination. Barrel has a microfinish for precision fit and the tips have the accuracy to provide required drainage rate. Each burette has a durable colored scale and dust cover.
£245.24 - £288.97GBP / Case of 1
£294.29 - £346.76GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Tolerance
Availability Pricing
Pyrex 2094-250 2094 Large Volume Class B Glass Burette, 250 mL; 1/Cs WZ-34542-30 Mfr # 2094-250
COMPARE
250 2
£245.24GBP / Case of 1
£294.29GBP (incl VAT)
Pyrex 2094-500 2094 Large Volume Class B Glass Burette, 500 mL; 1/Cs WZ-34542-32 Mfr # 2094-500
COMPARE
500 2.5
£249.25GBP / Case of 1
£299.10GBP (incl VAT)
Pyrex 2094-1L 2094 Large Volume Class B Glass Burette, 1000 mL; 1/Cs WZ-34542-34 Mfr # 2094-1L
COMPARE
1000 5
£288.97GBP / Case of 1
£346.76GBP (incl VAT)