This product is now discontinued, View Related Products

Hamilton PRP-X200 Strong Cation Exchange Columns

Efficient separations with durable media
Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene)
stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-X200 Strong Cation Exchange Columns contain poly (styrene-divinylbenzene) sulfonate. These columns also offer superior, rapid resolution; exchange capacity is 0.035 mEq/g.
£755.47 - £813.59GBP / Each
£906.56 - £976.31GBP (incl VAT)

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Column Particle Size (µm) Description
Availability Pricing
Hamilton 79441 PRP-X200 Cation Exchange Column, Fast Analytical, 150x4.1mm ID; 10um WZ-01943-00 Mfr # 79441
COMPARE
10 PRP-X200 Cation Exchange Column, Fast Analytical, 150x4.1mm ID; 10um
£755.47GBP / Each
£906.56GBP (incl VAT)
Hamilton 79442 PRP-X200 Cation Exchange Column, Analytical, 250x4.1mm ID; 10um WZ-01943-02 Mfr # 79442
COMPARE
10 PRP-X200 Cation Exchange Column, Analytical, 250x4.1mm ID; 10um
£813.59GBP / Each
£976.31GBP (incl VAT)
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat