Hamilton® PRP-X100 Strong Anion Exchange Columns
Efficient separations with durable media
  • High selectivity, efficiency, and capacity
Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene) stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-X100 Strong Anion Exchange Columns contain poly styrene-divinylbenzene)-trimethyl ammonium. These columns offer superior, rapid resolution. Exchange capacity is 0.2 mEq/g.
£784.53 - £848.65GBP / Each
£941.44 - £1,018.38GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Hamilton PRP-X100 Anion Exchange Column, Fast Analytical, 150x4.1mm ID; 10um WZ-01942-02 Mfr # 79434
COMPARE
4.1 150 10
£784.53GBP / Each
£941.44GBP (incl VAT)
Hamilton PRP-X100 Anion Exchange Column, Analytical, 250x4.1mm ID; 10um WZ-01942-04 Mfr # 79433
COMPARE
4.1 250 10
£848.65GBP / Each
£1,018.38GBP (incl VAT)