Winters Instruments Pressure System Tridicators
Measure both pressure and temperature using one gauge
  • 2.5” (63mm) and 3” (75mm) dials
  • Dual scale (°F & °C)
  • Back or bottom connection
  • Brass wetted parts
  • 1” (25mm), 2” (50mm) and 2.5” (63mm) stems
  • Multiple pressure and temperature ranges available
  • Can be used with TST thermowells
  • Ranges are compliant to industry standard ASME Boiler Code Section IV HG-612
  • 5 year warranty
Dual-use tridicators are specially constructed to indicate both pressure and temperature in one compact instrument. Applications include boilers, heating systems, or any other hot water systems that require measurement of pressure and temperature. Gauge pressure and temperature is indicated in psi and °F.
£29.91 - £50.37GBP / Each
£35.89 - £60.44GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Process Connection
Availability Pricing
Winters Instruments Pressure System Tridicator 0 to 75 psi / 70 to 320°F WZ-68618-02 Mfr # TTD401
COMPARE
75 1/4\" NPT, back
£29.91GBP / Each
£35.89GBP (incl VAT)
Winters Instruments Pressure System Tridicator 0 to 200 psi / 70 to 320°F WZ-68618-12 Mfr # TTD407
COMPARE
200 1/2\" NPT, back
£50.37GBP / Each
£60.44GBP (incl VAT)