Precision PTFE Plug Valve
Designed for caustic and high-purity use
Plug valves are manufactured from all PTFE with PVDF stem nuts. The PTFE body and stem are designed for use with harsh chemicals as well as ultra clean DI water.
The construction is a straight flow pattern with a tapered body that matches the tapered plug for positive shut-off. Various orifice and port sizes are available as standard items. Other sizes and configurations are available on request.
£291.00GBP / Each
£349.20GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Inlet Port Orifice (in)
Availability Pricing
COMPARE
NPT(F) 1/4" NPT(F) 0.25
£291.00GBP / Each
£349.20GBP (incl VAT)
PTFE Plug Valve with 1/4" Tube Compression Connections, 0.188" Orifice, 6-turn WZ-98517-16 Mfr # PPV-43T-A
COMPARE
Compression 1/4" compression 0.188
£291.00GBP / Each
£349.20GBP (incl VAT)