This product is now discontinued, View Related Products

Plum Emergency Personal Eye and Skin Washes

Flexible, form-fitting eye cup ensures gentle and effective rinsing
  • Bendable eye cup keeps eyelid open during use
  • All solutions meet U.S. Federal Regulations, comply with ANSI standards, and are FDA compliant for personal eyewash
Ideal for emergency rinsing of debris, and neutralization of acids or alkalis in the eyes. Bottles with a green eye cup are sterile saline eye wash; use to wash particulate debris from the eye. Bottles with blue eye cups contain a sterile phosphate buffer solution; use to rapidly neutralize acids, alkalis, and other chemicals in the eye.
To use simply twist the cap to open, apply the eye cup to the affected eye, and squeeze the bottle. Control the flow of solution with hand pressure on the bottle for more efficient rinsing and less waste. Each bottle of DUO eyewash releases two streams to rinse both eyes at once.

Compliance: ANSI Z358.1-2004
Call For Price

19 variations of this product are available.

See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Scienceware F24880-0023 Plum Sterile Saline Eyewash Refill, 200 mL, 3 bottles/pack WZ-06796-30 Mfr # F24880-0023
COMPARE
Plum Sterile Saline Eyewash Refill, 200 mL, 3 bottles/pack
Call For Price
Scienceware F24880-0053 Plum Sterile Saline Eyewash Refill, 500 mL, 3 bottles/pack WZ-06796-31 Mfr # F24880-0053
COMPARE
Plum Sterile Saline Eyewash Refill, 500 mL, 3 bottles/pack
Call For Price
Scienceware Plum Sterile Saline Eyewash Station, 500 mL WZ-06796-32 Mfr # F24880-4001
COMPARE
Plum Sterile Saline Eyewash Station, 500 mL
Call For Price
Scienceware F24880-4002 Plum Double Sterile Saline Eyewash Station, two 500 mL bottles WZ-06796-33 Mfr # F24880-4002
COMPARE
Plum Double Sterile Saline Eyewash Station, two 500 mL bottles
Call For Price
Scienceware F24880-5002 Plum Covered Double Saline Eyewash Station, 2 x 500 mL WZ-06796-34 Mfr # F24880-5002
COMPARE
Plum Covered Double Saline Eyewash Station, 2 x 500 mL
Call For Price
Scienceware F24880-1023 Plum Sterile pH Neutralizing Eyewash Refill, 200 mL, 3 bottles/pack WZ-06796-35 Mfr # F24880-1023
COMPARE
Plum Sterile pH Neutralizing Eyewash Refill, 200 mL, 3 bottles/pack
Call For Price
Scienceware F24880-1103 Plum Sterile pH Neutralizing Eyewash Refill, 1000 mL, 3 bottles/pack WZ-06796-36 Mfr # F24880-1103
COMPARE
Plum Sterile pH Neutralizing Eyewash Refill, 1000 mL, 3 bottles/pack
Call For Price
Scienceware F24880-4003 Plum Saline/pH Neutralizing Eyewash Station, 500 x 200 mL WZ-06796-37 Mfr # F24880-4003
COMPARE
Plum Saline/pH Neutralizing Eyewash Station, 500 x 200 mL
Call For Price
Scienceware F24880-5003 Plum Saline/pH Neutralizing Eyewash Station, 50 x 200 mL WZ-06796-38 Mfr # F24880-5003
COMPARE
Plum Saline/pH Neutralizing Eyewash Station, 50 x 200 mL
Call For Price
Scienceware Plum Sterile pH Neutralizing Emergency Shower Station, 1000 mL WZ-06796-39 Mfr # F24880-2100
COMPARE
Plum Sterile pH Neutralizing Emergency Shower Station, 1000 mL
Call For Price
Scienceware Plum Sterile Saline Eyewash Station, 1000 mL WZ-06796-40 Mfr # F24880-4010
COMPARE
Plum Sterile Saline Eyewash Station, 1000 mL
Call For Price
Scienceware Plum Double Sterile Saline Eyewash Station, two 1000 mL bottles WZ-06796-41 Mfr # F24880-4012
COMPARE
Plum Double Sterile Saline Eyewash Station, two 1000 mL bottles
Call For Price
Scienceware F24880-8010 Plum Sterile Saline DUO Eyewash Station, 1000 mL WZ-06796-42 Mfr # F24880-8010
COMPARE
Plum Sterile Saline DUO Eyewash Station, 1000 mL
Call For Price
Scienceware F24880-8502 Plum DUO Double Saline Eyewash Station, 2 x 500 mL WZ-06796-43 Mfr # F24880-8502
COMPARE
Plum DUO Double Saline Eyewash Station, 2 x 500 mL
Call For Price
Scienceware F24880-8403 Plum Duo Saline/pH Neutralizing Eyewash Station, 1000 x 500 mL WZ-06796-44 Mfr # F24880-8403
COMPARE
Plum Duo Saline/pH Neutralizing Eyewash Station, 1000 x 500 mL
Call For Price
Scienceware F24880-8503 Plum Duo Covered Saline/pH Neutralizing Eyewash Station, 1000 x 500 mL WZ-06796-45 Mfr # F24880-8503
COMPARE
Plum Duo Covered Saline/pH Neutralizing Eyewash Station, 1000 x 500 mL
Call For Price
Scienceware F24880-0103 Plum Sterile Saline Eyewash Refill, 1000 mL, 3 bottles/pack WZ-06796-46 Mfr # F24880-0103
COMPARE
Plum Sterile Saline Eyewash Refill, 1000 mL, 3 bottles/pack
Call For Price
Scienceware F24880-8103 Plum DUO Sterile Saline Eyewash Refill, 1000 mL, 6 bottles/pack WZ-06796-47 Mfr # F24880-8103
COMPARE
Plum DUO Sterile Saline Eyewash Refill, 1000 mL, 6 bottles/pack
Call For Price
Scienceware F24880-8023 Plum DUO Sterile pH Neutralizing Eyewash Refill, 500 mL; 6/Pk WZ-06796-48 Mfr # F24880-8023
COMPARE
Plum DUO Sterile pH Neutralizing Eyewash Refill, 500 mL; 6/Pk
Call For Price
Showing 1- 19 of 19

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat