This product is now discontinued, View Related Products
Phenix Portable Hipot Megohmmeters
FEATURES
  • Integral megohmmeter measures insulation resistance up to 500,000 megohms at 2.5 KVDC or up to 1,000,000 megohms @ 5.0 KVDC
  • Voltage output continuously adjustable up to 5,000 VAC or 6,000 VDC
  • “Zero Start” voltage interlock
  • Large, easy-to-read analog meters
  • Leakage current measurements from 0.1 to 5,000 microamps DC and from 1.0 to 5.0 milliamps AC
  • Adjustable overcurrent trip point
  • Rotary switch selection simplifies operation
  • Lightweight, portable and enclosed in a rugged, fiberglass carrying case
  • Model PM6 has same specifications as PAD56, less AC output capability
Call For Price
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
AC Current Accuracy AC Current Range (mA) AC Current Resolution (mA)
Availability Pricing
COMPARE
Call For Price
Phenix Technologies PAD56 Portable AC/DC HiPot Megohmeter WZ-20023-40 Mfr # PAD56
COMPARE
Call For Price
Phenix Technologies PM15-2 15k VDC Hipot-Megohmeter, 120V AC WZ-20023-55 Mfr # PM15-2