Pentair In-line Cartridge Filtration Systems
A complete filtration solution in one unit
  • Include a pre- and post- filter
  • A variety of sizes to meet your capacity needs
  • Ideal choice for post-RO, under-sink, ice-maker, and food service applications
Each in-line filter includes an injection-molded polypropylene body and cap, and pre- and post-filters. Choose from either FPT connections or John Guest® push-to-connect fittings. Two unique filter types are available — for the removal of chlorine taste and odor, or removal of rust and sediment. All filters have a maximum temperature rating of 100°F (38°C) and 125 psi pressure rating. Filters meet NSF standards.
£13.53 - £29.85GBP / Each
£16.24 - £35.82GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Flow Rate Port Size Description
Availability Pricing
Pentair 255576-43 In-Line Filtration System for Sediment and Rust, 5 µm, 4 LPM, 3/8" NPT(F) WZ-01507-63 Mfr # 255576-43
COMPARE
4 LPM (1 GPM) 3/8" NPT(F) In-Line Filtration System for Sediment and Rust, 5 µm, 4 LPM, 3/8" NPT(F)
£13.53GBP / Each
£16.24GBP (incl VAT)
Pentair 255579-43 In-Line Filtration System for Sediment and Rust, 5 µm, 4 LPM, 1/4" John Guest WZ-01507-64 Mfr # 255579-43
COMPARE
4 LPM (1 GPM) 1/4" John Guest In-Line Filtration System for Sediment and Rust, 5 µm, 4 LPM, 1/4" John Guest
£16.70GBP / Each
£20.04GBP (incl VAT)
Pentair 255582-43 In-Line Filtration System for Sediment and Rust, 5 µm, 8 LPM, 1/4" John Guest WZ-01507-65 Mfr # 255582-43
COMPARE
8 LPM (2 GPM) 1/4" John Guest In-Line Filtration System for Sediment and Rust, 5 µm, 8 LPM, 1/4" John Guest
£22.39GBP / Each
£26.87GBP (incl VAT)
Pentair 255631-43 In-Line Filtration System for Chlorine Taste and Odor, 5 µm, 2 LPM, 1/4" NPT(F) WZ-01507-83 Mfr # 255631-43
COMPARE
2 LPM (0.5 GPM) 1/4" NPT(F) In-Line Filtration System for Chlorine Taste and Odor, 5 µm, 2 LPM, 1/4" NPT(F)
£25.88GBP / Each
£31.06GBP (incl VAT)
Pentair 255632-43 In-Line Filtration System for Chlorine Taste and Odor, 5 µm, 2 LPM, 3/8" NPT(F) WZ-01507-84 Mfr # 255632-43
COMPARE
2 LPM (0.5 GPM) 3/8"NPT(F) In-Line Filtration System for Chlorine Taste and Odor, 5 µm, 2 LPM, 3/8" NPT(F)
£25.88GBP / Each
£31.06GBP (incl VAT)
Pentair 255635-43 In-Line Filtration System for Chlorine Taste and Odor, 5 µm, 2 LPM, 1/4" John Guest WZ-01507-85 Mfr # 255635-43
COMPARE
2 LPM (0.5 GPM) 1/4" John Guest In-Line Filtration System for Chlorine Taste and Odor, 5 µm, 2 LPM, 1/4" John Guest
£29.39GBP / Each
£35.27GBP (incl VAT)
Pentair 255636-43 In-Line Filtration System for Chlorine Taste and Odor, 5 µm, 2 LPM, 3/8" John Guest WZ-01507-86 Mfr # 255636-43
COMPARE
2 LPM (0.5 GPM) 3/8" John Guest In-Line Filtration System for Chlorine Taste and Odor, 5 µm, 2 LPM, 3/8" John Guest
£29.85GBP / Each
£35.82GBP (incl VAT)
Showing 1- 7 of 7