IDEX® PEEK Low-Pressure Chromatography Fitting Kit
Save time and money with a fitting kits
  • Constructed of chemically inert PEEK plastic
PEEK Low-Pressure Unions, Tees, and Crosses hold up to 1000 psi (1/16" tubing), or to 500 psi (1/8" tubing). PEEK flangeless fittings included. Models 02008-00, -10, and -20 take 1/16" OD tubing; models 02008-04, -14, and -24 take 1/8" OD tubing.
£533.52GBP / Set of 1
£640.22GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Description
Availability Pricing
Idex Low Pressure Chromatography Fittings Kit WZ-02008-52 Mfr # 1456
COMPARE
PEEK HPLC/Chromatography Low Pressure Chromatorgraphy Fittings Kit
£533.52GBP / Set of 1
£640.22GBP (incl VAT)