PEAK Scientific Precision Hydrogen SL Generators
The smallest hydrogen generator for GC-FID
  • 99.9995% purity at 100 psi output pressure
  • Far safer than highly pressurized cylinders
  • Produces hydrogen at up to 200 cc/min with no compromise on safety
  • Easy one-button startup and shutdown
  • Models available with auto-water fill or pressurized water fill
The Precision Hydrogen SL line has been developed to offer a streamlined laboratory-grade hydrogen gas solution for your GC flame detectors. Units are safe, simple to use, easy to maintain, and take up minimal bench space.
Hydrogen in the lab is always a source of concern – high pressure cylinders can be a risk to lab personnel, as well as a large volume of gas that can leak into the lab environment. With Precision Hydrogen SL in your lab, you eliminate these risks entirely. The generator produces only gas on-demand to meet instrument needs and stores less than 150 mL of gas in the instrument, with an automatic fail-safe to shut off the generator in the event of any leak.
The generator is available with manual, automatic, or pressurized water fill, and in a choice of black or white housing.
£7,452.61 - £8,921.85GBP / Each
£8,943.13 - £10,706.22GBP (incl VAT)
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Power (VAC) Description
Availability Pricing
PEAK Scientific Precision Hydrogen SL Generator, 100 cc, Manual Fill, Black; 110/240 VAC WZ-86443-16 Mfr # 3302946
COMPARE
100 110 / 240 Generator, 100 cc, Manual Fill, Black; 110/240 VAC
£7,452.61GBP / Each
£8,943.13GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 100 cc, Manual Fill, White; 110/240 VAC WZ-86443-17 Mfr # 3302943
COMPARE
100 110 / 240 Generator, 100 cc, Manual Fill, White; 110/240 VAC
£7,452.61GBP / Each
£8,943.13GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 100 cc, Auto Fill, Black; 110/240 VAC WZ-86443-18 Mfr # 3302947
COMPARE
100 110 / 240 Generator, 100 cc, Auto Fill, Black; 110/240 VAC
£7,792.25GBP / Each
£9,350.70GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 100 cc, Auto Fill, White; 110/240 VAC WZ-86443-19 Mfr # 3302944
COMPARE
100 110 / 240 Generator, 100 cc, Auto Fill, White; 110/240 VAC
£7,792.25GBP / Each
£9,350.70GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 100 cc, Pressurized Fill, Black; 110/240 VAC WZ-86443-20 Mfr # 3302948
COMPARE
100 110 / 240 Generator, 100 cc, Pressurized Fill, Black; 110/240 VAC
£8,131.90GBP / Each
£9,758.28GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 100 cc, Pressurized Fill, White; 110/240 VAC WZ-86443-21 Mfr # 3302945
COMPARE
100 110 / 240 Generator, 100 cc, Pressurized Fill, White; 110/240 VAC
£8,131.90GBP / Each
£9,758.28GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 200 cc, Manual Fill, Black; 110/240 VAC WZ-86443-22 Mfr # 3302952
COMPARE
200 110 / 240 Generator, 200 cc, Manual Fill, Black; 110/240 VAC
£8,242.56GBP / Each
£9,891.07GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 200 cc, Manual Fill, White; 110/240 VAC WZ-86443-23 Mfr # 3302949
COMPARE
200 110 / 240 Generator, 200 cc, Manual Fill, White; 110/240 VAC
£8,242.56GBP / Each
£9,891.07GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 200 cc, Auto Fill, Black; 110/240 VAC WZ-86443-24 Mfr # 3302953
COMPARE
200 110 / 240 Generator, 200 cc, Auto Fill, Black; 110/240 VAC
£8,583.05GBP / Each
£10,299.66GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 200 cc, Auto Fill, White; 110/240 VAC WZ-86443-25 Mfr # 3302950
COMPARE
200 110 / 240 Generator, 200 cc, Auto Fill, White; 110/240 VAC
£8,583.05GBP / Each
£10,299.66GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 200 cc, Pressurized Fill, Black; 110/240 VAC WZ-86443-26 Mfr # 3302954
COMPARE
200 110 / 240 Generator, 200 cc, Pressurized Fill, Black; 110/240 VAC
£8,921.85GBP / Each
£10,706.22GBP (incl VAT)
PEAK Scientific Precision Hydrogen SL Generator, 200 cc, Pressurized Fill, White; 110/240 VAC WZ-86443-27 Mfr # 3302951
COMPARE
200 110 / 240 Generator, 200 cc, Pressurized Fill, White; 110/240 VAC
£8,921.85GBP / Each
£10,706.22GBP (incl VAT)
Showing 1- 12 of 12