Parker Hannifin Union Fittings, Female NPT Threaded, Elbow
Chemically-inert PFA handles most aggressive chemicals
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings offer a more permanent installation. Choose the size to meet your application needs.
£32.64 - £56.34GBP / Each
£39.17 - £67.61GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Thread size NPT(F) (port 1) (in) Thread size NPT(F) (port 2) (in)
Availability Pricing
Parker Hannifin PE-2-P Fitting, PFA, Elbow, Threaded Union, 1/8" NPT(F) WZ-31321-08 Mfr # PE-2-P
COMPARE
PFA 1/8 1/8
£32.64GBP / Each
£39.17GBP (incl VAT)
Parker Hannifin PE-4-P Fitting, PFA, Elbow, Threaded Union, 1/4" NPT(F) WZ-31321-09 Mfr # PE-4-P
COMPARE
PFA 1/4 1/4
£39.73GBP / Each
£47.68GBP (incl VAT)
Parker Hannifin PE-6-P Fitting, PFA, Elbow, Threaded Union, 3/8" NPT(F) WZ-31321-10 Mfr # PE-6-P
COMPARE
PFA 3/8 3/8
£48.95GBP / Each
£58.74GBP (incl VAT)
Parker Hannifin PE-8-P Fitting, PFA, Elbow, Threaded Union, 1/2" NPT(F) WZ-31321-11 Mfr # PE-8-P
COMPARE
PFA 1/2 1/2
£56.34GBP / Each
£67.61GBP (incl VAT)