Parker Hannifin Plug Fittings, Male NPT Threaded, Straight
Deigned for permanent connection
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 200 psi at 77ºF ( 25ºC). Usable from -65 to 190ºF (-53 to 87ºC).
£13.69 - £31.02GBP / Each
£16.43 - £37.22GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Connection Type Thread size NPT(M) (port 1) (in)
Availability Pricing
Parker Hannifin Fitting, PFA, Straight, Threaded Plug, 1/8" NPT(M) WZ-31321-13 Mfr # P-2-P
COMPARE
PFA NPT(M) 1/8
£17.23GBP / Each
£20.68GBP (incl VAT)
Parker Hannifin Fitting, PFA, Straight, Threaded Plug, 1/4"NPT(M) WZ-31321-14 Mfr # P-4-P
COMPARE
PFA NPT(M) 1/4
£19.58GBP / Each
£23.50GBP (incl VAT)
Parker Hannifin Fitting, PFA, Straight, Threaded Plug, 3/8" male NPT fitting WZ-31321-15 Mfr # P-6-P
COMPARE
PFA NPT(M) 3/8
£20.69GBP / Each
£24.83GBP (incl VAT)
Parker Hannifin Fitting, PFA, Straight, Threaded Plug, 1/2" NPT(M) WZ-31321-16 Mfr # P-8-P
COMPARE
PFA NPT(M) 1/2
£24.78GBP / Each
£29.74GBP (incl VAT)
Parker Hannifin Fitting, PFA, Straight, Threaded Plug, 3/4" NPT(M) WZ-31321-17 Mfr # P-12-P
COMPARE
PFA NPT(M) 3/4
£31.02GBP / Each
£37.22GBP (incl VAT)
Parker Hannifin Fitting, 316 Stainless Steel, Straight, Threaded Hex Plug, 1/4" NPT(M), 9/16" Hex WZ-31820-18 Mfr # 4 PH-SS
COMPARE
316 Stainless Steel NPT(M) 1/4
£13.69GBP / Each
£16.43GBP (incl VAT)
Parker Hannifin Fitting, 316 Stainless Steel, Straight, Threaded Hex Plug, 1/2" NPT(M), 7/8" Hex WZ-31820-19 Mfr # 8 PH-SS
COMPARE
316 Stainless Steel NPT(M) 1/2
£24.57GBP / Each
£29.48GBP (incl VAT)
Showing 1- 7 of 7