Parker Hannifin Adapter Fittings, Compression to Male Threaded, Straight
Excellent choice to connect rigid tubing systems
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
£16.01 - £212.36GBP / Each
£19.21 - £254.83GBP (incl VAT)
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Thread size NPT(M) (port 2) (in)
Availability Pricing
Parker Hannifin P4MC2 Fitting, Black Polypropylene, Compression to Thread, 1/4" OD x 1/8" NPT(M); 25/Pk WZ-06444-00 Mfr # P4MC2
COMPARE
1/4 Polypropylene 1/8
£63.92GBP / Pkg of 25
£76.70GBP (incl VAT)
Parker Hannifin Fitting, Black Polypropylene, Compression to Thread, 1/4" OD x 1/4" NPT(M); 25/Pk WZ-06444-01 Mfr # P4MC4
COMPARE
1/4 Polypropylene 1/4
£61.76GBP / Pkg of 25
£74.11GBP (incl VAT)
Parker Hannifin Fitting, Black Polypropylene, Compression to Thread, 3/8" OD x 3/8" NPT(M); 25/Pk WZ-06444-02 Mfr # P6MC6
COMPARE
3/8 Polypropylene 3/8
£76.93GBP / Pkg of 25
£92.32GBP (incl VAT)
Parker Hannifin Straight male pipe Adapter, Black PP, 3/8" OD x 1/2" NPT, 1-3/4" x 1" x 1/4" WZ-06444-03 Mfr # P6MC8
COMPARE
3/8 Polypropylene 1/2
£91.01GBP / Pkg of 25
£109.21GBP (incl VAT)
Parker Hannifin Straight male pipe Adapter, Black PP, 1/2" OD x 1/2" NPT, 1-7/8" x 1" x 3/8" WZ-06444-04 Mfr # P8MC8
COMPARE
1/2 Polypropylene 1/2
£117.01GBP / Pkg of 25
£140.41GBP (incl VAT)
Parker Hannifin Straight male pipe adapter, Black PP, 5/8" OD x 1/2" NPT, 2" x 1-1/8" x 1/2" WZ-06444-06 Mfr # P10MC8
COMPARE
5/8 Polypropylene 1/2
£136.52GBP / Pkg of 25
£163.82GBP (incl VAT)
Parker Hannifin Straight male pipe Adapter, Black PP, 5/8" OD x 3/4" NPT, 2" x 1-1/8" x 1/2" WZ-06444-07 Mfr # P10MC12
COMPARE
5/8 Polypropylene 3/4
£141.93GBP / Pkg of 25
£170.32GBP (incl VAT)
Parker Hannifin Fitting, Black Polypropylene, Straight, Compression to Threaded Adapter, 1/4" OD x 1/8" NPT(M); 25/PK WZ-06444-10 Mfr # P4FC2
COMPARE
1/4 Polypropylene 1/8
£212.36GBP / Pkg of 25
£254.83GBP (incl VAT)
Parker Hannifin 1MSCN1N-316 Compression pipe adapter, 1/16" x 1/16", Stainless Steel, Each WZ-30486-34 Mfr # 1MSCN1N-316
COMPARE
1/16 Stainless Steel 1/16
£28.86GBP / Each
£34.63GBP (incl VAT)
Parker Hannifin Compression pipe adapter, 1/8" x 1/8", Stainless Steel, Each WZ-30486-35 Mfr # 2MSCN2N-316
COMPARE
1/8 Stainless Steel 1/8
£17.40GBP / Each
£20.88GBP (incl VAT)
Parker Hannifin Compression pipe adapter, 1/4" x 1/8", Stainless Steel, Each WZ-30486-36 Mfr # 4MSCN2N-316
COMPARE
1/4 Stainless Steel 1/8
£16.01GBP / Each
£19.21GBP (incl VAT)
Parker Hannifin Compression pipe adapter, 1/4" x 1/4", Stainless Steel, Each WZ-30486-37 Mfr # 4MSC4N-316
COMPARE
1/4 Stainless Steel 1/4
£17.59GBP / Each
£21.11GBP (incl VAT)
Parker Hannifin Compression pipe adapter, 1/4" x 1/2", Stainless Steel, Each WZ-30486-38 Mfr # 4MSCN8N-316
COMPARE
1/4 Stainless Steel 1/2
£28.86GBP / Each
£34.63GBP (incl VAT)
Parker Hannifin 6MSCN4N-316 Compression pipe adapter, 3/8" x 1/4", Stainless Steel, Each WZ-30486-40 Mfr # 6MSCN4N-316
COMPARE
3/8 Stainless Steel 1/4
£23.72GBP / Each
£28.46GBP (incl VAT)
Parker Hannifin 6MSCN6N-316 Compression pipe adapter, 3/8" x 3/8", Stainless Steel, Each WZ-30486-41 Mfr # 6MSCN6N-316
COMPARE
3/8 Stainless Steel 3/8
£26.49GBP / Each
£31.79GBP (incl VAT)
Parker Hannifin 6MSCN8N-316 Compression pipe adapter, 3/8" x 1/2", Stainless Steel, Each WZ-30486-42 Mfr # 6MSCN8N-316
COMPARE
3/8 Stainless Steel 1/2
£30.06GBP / Each
£36.07GBP (incl VAT)
Parker Hannifin 8MSCN4N-316 Compression pipe adapter, 1/2" x 1/4", Stainless Steel, Each WZ-30486-43 Mfr # 8MSCN4N-316
COMPARE
1/2 Stainless Steel 1/4
£32.50GBP / Each
£39.00GBP (incl VAT)
Parker Hannifin 8MSCN6N Compression pipe adapter, 1/2" x 3/8", Stainless Steel, Each WZ-30486-44 Mfr # 8MSCN6N
COMPARE
1/2 Stainless Steel 3/8
£33.59GBP / Each
£40.31GBP (incl VAT)
Parker Hannifin Compression pipe adapter, 1/2" x 1/2", Stainless Steel, Each WZ-30486-45 Mfr # 8MSCN8N-316
COMPARE
1/2 Stainless Steel 1/2
£34.67GBP / Each
£41.60GBP (incl VAT)
Parker Hannifin Compression pipe adapter, 5/8" x 3/8", Stainless Steel, Each WZ-30486-47 Mfr # 10MSCN6N-316
COMPARE
5/8 Stainless Steel 3/8
£37.92GBP / Each
£45.50GBP (incl VAT)
Showing 1 - 20 of 22