This product is now discontinued, View Related Products
Pacific Scientific OECO Gauss - Tesla Meters
Benchtop performance in a compact handheld design
  • Digital signal processor provides the latest measurement technology
  • Data Logging capability
  • 100X lower resolution (with low-field probe)
  • Frequency Response 2X better than competitor
  • The only handheld meter with DSP (Digital Signal Processing)
  • Auto Zero
  • Min/Max/Peak Hold
  • Auto Range
  • Relative Mode
  • Universal Serial Bus Interface
These autoranging meters measure in gauss and tesla, and feature auto zero, min/max and peak hold
readings, relative mode and built-in software for easy field calibration. The ergonomic design provides
comfort during measurements of magnetic fields around critical equipment.
Model 20001-29 also measures in ampere/meters and features an analog output and USB communication port for downloading measurements to your PC.
2 PRODUCT OPTIONS
Compareitem specifications
Range (Gauss) Resolution (Gauss)
Availability Pricing
Pacific Scientific-OECO 5170 Gaussmeter w/4in Probe AC Adapter Hard Carrying Case WZ-20001-28 Mfr # 5170
COMPARE
200 G; 2, 20 kG 0.1, 1, 10 G
Call For Price
Pacific Scientific-OECO 5180 Gaussmeter w/4in Probe AC Adapter Hard Carrying Case WZ-20001-29 Mfr # 5180 WITH STD18-0404 TRANSVERSE PROBE INCLUDED
COMPARE
300 G; 3, 30 kG 0.1, 1, 10 G