This product is now discontinued, View Related Products

Omnifit® SolventPlus™ Chromatography Columns with Two Fixed Endpieces

Get the highest chemical compatibility
  • Exceptional chemical resistance with wetted parts of PTFE, ETFE, borosilicate glass, and Chemraz®
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
These SolventPlus columns are designed for techniques using organic solvents such as reversed phase chromatography, but are equally compatible with aqueous solvent systems. The most inert materials are present in the fluid path to provide superior chemical resistance. Use the columns in low pressure or gravity feed applications.

Columns come with two fixed endpieces pre-assembled with 30 µm PTFE frits. Modular design allow you to mix and match endpieces to suit your application. Each column is supplied assembled and ready to use, with all the tubing, fittings, and connections needed to get started.
Call For Price

26 variations of this product are available.

See All Product Options Below
26 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Column Inside Diameter (ID) (mm) Column Length (mm)
Availability Pricing
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 50 mm; 1/ea WZ-11946-02 Mfr # 006SCC-06-05-FF
COMPARE
0.7 6.6 50
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 100 mm; 1/ea WZ-11946-05 Mfr # 006SCC-06-10-FF
COMPARE
2.4 6.6 100
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 150 mm; 1/ea WZ-11946-08 Mfr # 006SCC-06-15-FF
COMPARE
4.1 6.6 150
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 250 mm; 1/ea WZ-11946-11 Mfr # 006SCC-06-25-FF
COMPARE
7.5 6.6 250
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 6.6 x 400 mm; 1/ea WZ-11946-14 Mfr # 006SCC-06-40-FF
COMPARE
12.7 6.6 400
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 10 x 100 mm; 1/ea WZ-11946-17 Mfr # 006SCC-10-10-FF
COMPARE
5.5 10 100
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 10 x 150 mm; 1/ea WZ-11946-20 Mfr # 006SCC-10-15-FF
COMPARE
9.4 10 150
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 10 x 250 mm; 1/ea WZ-11946-23 Mfr # 006SCC-10-25-FF
COMPARE
17.3 10 250
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 10 x 400 mm; 1/ea WZ-11946-26 Mfr # 006SCC-10-40-FF
COMPARE
29.1 10 400
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 10 x 500 mm; 1/ea WZ-11946-29 Mfr # 006SCC-10-50-FF
COMPARE
36.9 10 500
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 15 x 100 mm; 1/ea WZ-11946-32 Mfr # 006SCC-15-10-FF
COMPARE
12.4 15 100
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 15 x 150 mm; 1/ea WZ-11946-35 Mfr # 006SCC-15-15-FF
COMPARE
21.2 15 150
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 15 x 250 mm; 1/ea WZ-11946-38 Mfr # 006SCC-15-25-FF
COMPARE
38.9 15 250
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 15 x 400 mm; 1/ea WZ-11946-41 Mfr # 006SCC-15-40-FF
COMPARE
65.4 15 400
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 15 x 500 mm; 1/ea WZ-11946-44 Mfr # 006SCC-15-50-FF
COMPARE
83 15 500
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 25 x 100 mm; 1/ea WZ-11946-47 Mfr # 006SCC-25-10-FF
COMPARE
34.4 25 100
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 25 x 150 mm; 1/ea WZ-11946-50 Mfr # 006SCC-25-15-FF
COMPARE
58.9 25 150
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 25 x 250 mm; 1/ea WZ-11946-53 Mfr # 006SCC-25-25-FF
COMPARE
108 25 250
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 25 x 400 mm; 1/ea WZ-11946-56 Mfr # 006SCC-25-40-FF
COMPARE
181.6 25 400
Call For Price
Diba Omnifit® SolventPlus™ Chromatography Column w/ 2 Fixed Endpieces, 25 x 500 mm; 1/ea WZ-11946-59 Mfr # 006SCC-25-50-FF
COMPARE
230.7 25 500
Call For Price
Showing 1 - 20 of 26

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat