This product is now discontinued, View Related Products

Omnifit® Replacement Glass Tubes for BenchMark™, HiT™, and SolventPlus™ Columns

Mix and match column length while using your existing endpieces for ultimate flexibility
Use a different length tube with your existing endpieces to meet your specific requirements.
Call For Price

26 variations of this product are available.

See All Product Options Below
26 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm)
Availability Pricing
Diba Omnifit® Glass Column Only, 6.6 x 50 mm; 1/ea WZ-11945-68 Mfr # 006RG-06-05
COMPARE
6.6 50
Call For Price
Diba Omnifit® Glass Column Only, 6.6 x 100 mm; 1/ea WZ-11945-69 Mfr # 006RG-06-10
COMPARE
6.6 100
Call For Price
Diba Omnifit® Glass Column Only, 6.6 x 150 mm; 1/ea WZ-11945-70 Mfr # 006RG-06-15
COMPARE
6.6 150
Call For Price
Diba Omnifit® Glass Column Only, 6.6 x 250 mm; 1/ea WZ-11945-71 Mfr # 006RG-06-25
COMPARE
6.6 250
Call For Price
Diba Omnifit® Glass Column Only, 6.6 x 400 mm; 1/ea WZ-11945-72 Mfr # 006RG-06-40
COMPARE
6.6 400
Call For Price
Diba Omnifit® Glass Column Only, 10 x 100 mm; 1/ea WZ-11945-73 Mfr # 006RG-10-10
COMPARE
10 100
Call For Price
Diba Omnifit® Glass Column Only, 10 x 150 mm; 1/ea WZ-11945-74 Mfr # 006RG-10-15
COMPARE
10 150
Call For Price
Diba Omnifit® Glass Column Only, 10 x 250 mm; 1/ea WZ-11945-75 Mfr # 006RG-10-25
COMPARE
10 250
Call For Price
Diba Omnifit® Glass Column Only, 10 x 400 mm; 1/ea WZ-11945-76 Mfr # 006RG-10-40
COMPARE
10 400
Call For Price
Diba Omnifit® Glass Column Only, 10 x 500 mm; 1/ea WZ-11945-77 Mfr # 006RG-10-50
COMPARE
10 500
Call For Price
Diba Omnifit® Glass Column Only, 15 x 100 mm; 1/ea WZ-11945-78 Mfr # 006RG-15-10
COMPARE
15 100
Call For Price
Diba Omnifit® Glass Column Only, 15 x 150 mm; 1/ea WZ-11945-79 Mfr # 006RG-15-15
COMPARE
15 150
Call For Price
Diba Omnifit® Glass Column Only, 15 x 250 mm; 1/ea WZ-11945-80 Mfr # 006RG-15-25
COMPARE
15 250
Call For Price
Diba Omnifit® Glass Column Only, 15 x 400 mm; 1/ea WZ-11945-81 Mfr # 006RG-15-40
COMPARE
15 400
Call For Price
Diba Omnifit® Glass Column Only, 15 x 500 mm; 1/ea WZ-11945-82 Mfr # 006RG-15-50
COMPARE
15 500
Call For Price
Diba Omnifit® Glass Column Only, 25 x 100 mm; 1/ea WZ-11945-83 Mfr # 006RG-25-10
COMPARE
25 100
Call For Price
Diba Omnifit® Glass Column Only, 25 x 150 mm; 1/ea WZ-11945-84 Mfr # 006RG-25-15
COMPARE
25 150
Call For Price
Diba Omnifit® Glass Column Only, 25 x 250 mm; 1/ea WZ-11945-85 Mfr # 006RG-25-25
COMPARE
25 250
Call For Price
Diba Omnifit® Glass Column Only, 25 x 400 mm; 1/ea WZ-11945-86 Mfr # 006RG-25-40
COMPARE
25 400
Call For Price
Diba Omnifit® Glass Column Only, 25 x 500 mm; 1/ea WZ-11945-87 Mfr # 006RG-25-50
COMPARE
25 500
Call For Price
Showing 1 - 20 of 26

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat