This product is now discontinued, View Related Products

Omegadyne Tension and Compression Load Cells - Stainless Steel Cantilever Beams

Stainless steel cantilever beams
  • FM Intrinsically Safe
  • All Stainless Steel for Harsh Industrial Applications
  • 0.25% Interchangeability for Multiple Load Cell Applications
  • 5-Point Calibration Provided
Call For Price

9 variations of this product are available.

See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs)
Availability Pricing
Omegadyne LC501-100 SS Cantilever Beam, Cable Style; 100 lb WZ-59890-33 Mfr # LC501-100
COMPARE
100
Call For Price
Omegadyne LC501-10K SS Cantilever Beam, Cable Style; 10,000 lb WZ-59890-34 Mfr # LC501-10K
COMPARE
10000
Call For Price
Omegadyne LC501-1K SS Cantilever Beam, Cable Style; 1000 lb WZ-59890-35 Mfr # LC501-1K
COMPARE
1000
Call For Price
Omegadyne LC501-200 SS Cantilever Beam, Cable Style; 200 lb WZ-59890-36 Mfr # LC501-200
COMPARE
200
Call For Price
Omegadyne LC501-2K SS Cantilever Beam, Cable Style; 2000 lb WZ-59890-37 Mfr # LC501-2K
COMPARE
2000
Call For Price
Omegadyne LC501-500 SS Cantilever Beam, Cable Style; 500 lb WZ-59890-38 Mfr # LC501-500
COMPARE
500
Call For Price
Omegadyne LC501-5K SS Cantilever Beam, Cable Style; 5000 lb WZ-59890-39 Mfr # LC501-5K
COMPARE
5000
Call For Price
Omegadyne LC501-100K Cantilever Beam Tension/Compression Load Cell; 100,000 lb WZ-93955-40 Mfr # LC501-100K
COMPARE
100000
Call For Price
Omegadyne LC501-50K Cantilever Beam Tension/Compression Load Cell; 30,000 lb WZ-93955-41 Mfr # LC501-30K
COMPARE
50000
Call For Price
Showing 1- 9 of 9

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat