This product is now discontinued, View Related Products
Ohaus YJ Series Jewelry Balances
Portable and economical—ideal for weighing fine stones, gems, and precious metals
  • Bright blue backlit LCD makes viewing results easy in dim lighting
  • Hinged draft shield cover ensures exact results
  • Automatic shutoff conserves battery life
  • Adjustable feet and leveling bubble provide quick setup
These portable electronic balances are designed to meet the highest quality standards and weighing needs for the jewelry industry. The carat models are equipped for precise weighing of fine stones and gems, while the gold models are best suited for weighing gold, precious metals and other fine jewelry. All units feature simple-to-use four-button operation and low-battery indicator.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Readability (g) Description
Availability Pricing
Ohaus YJ103 Carat Series Jewelry Balance, 100 ct x 0.005 ct WZ-11335-77 Mfr # YJ103
COMPARE
20 0.001 Carat Series Jewelry Balance, 100 ct x 0.005 ct
Call For Price
Ohaus YJ152 Carat Series Jewelry Balance, 150 ct x 0.01 ct WZ-11335-78 Mfr # YJ152
COMPARE
30 0.002 Carat Series Jewelry Balance, 150 ct x 0.01 ct
Call For Price
Ohaus YJ102 Gold Series Jewelry Balance, 100 g x 0.01 g WZ-11335-79 Mfr # YJ102
COMPARE
100 0.01 Gold Series Jewelry Balance, 100 g x 0.01 g
Call For Price
Ohaus YJ301 Gold Series Jewelry Balance, 300 g x 0.1 g WZ-11335-80 Mfr # YJ301
COMPARE
300 0.1 Gold Series Jewelry Balance, 300 g x 0.1 g
Call For Price
Ohaus YJ501 Gold Series Jewelry Balance, 500 g x 0.1 g WZ-11335-81 Mfr # YJ501
COMPARE
500 0.1 Gold Series Jewelry Balance, 500 g x 0.1 g