Ohaus Field-Test Beam Mechanical Balances
Accurately weigh a wide variety of heavy materials, even under the most adverse conditions
  • Large stabilized pan will hold oversized samples without tipping
  • Weighted base keeps the entire scale steady
  • Single sliding weight beam
  • Weights are stored within the base
  • Slotted weight set is included
£860.00 - £1,188.00GBP / Each
£1,032.00 - £1,425.60GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Readability (g) Description
Availability Pricing
Ohaus 2400-11 Field-Test Mechanical Balance, 16kg x 5g WZ-11010-73 Mfr # 2400-11
COMPARE
16000 5 Field-Test Mechanical Balance, 16kg x 5g
£860.00GBP / Each
£1,032.00GBP (incl VAT)
Ohaus 2400-12 Field-Test Mechanical Balance, 36 Lb x 0.01 Lb WZ-11010-74 Mfr # 2400-12
COMPARE
16329 4.5 Field-Test Mechanical Balance, 36 Lb x 0.01 Lb
£1,188.00GBP / Each
£1,425.60GBP (incl VAT)