Ohaus Defender Q-Series High Resolution Industrial Scale Base Only

Desighed for the industrial environment
  • Removable 304 stainless steel weighing pans offer quick cleaning
  • Black polyurethan painted steel frame
  • Durability, rigidity and structural integrity
  • IP-67 aluminum load cell
These durable rectangular bases feature welded tubular steel construction for industrial applications. Rugged, adjustable feet help prevent slippage on wet or uneven surfaces
Call For Price
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Readability (g)
Availability Pricing
Ohaus D10QR Defender Q-Series Standard Square Base 10kg 12" x 12" WZ-90453-03 Mfr # D10QR
COMPARE
10 25 1
Call For Price
Ohaus D25QR Defender Q-Series Standard Square Base 25kg 12" x 12" WZ-90453-04 Mfr # D25QR
COMPARE
25 50 2
Call For Price
Ohaus D50QL Defender Q-Series Standard Square Base 50kg 18" x 18" WZ-90453-05 Mfr # D50QL
COMPARE
50 100 5
Call For Price
Ohaus D100QL Defender Q-Series D50Ql Standard Square Base 100kg 18" x 18" WZ-90453-06 Mfr # D100QL
COMPARE
100 250 10
Call For Price
Ohaus D250QX Defender Standard Square Base 250kg 24" x 24" WZ-90453-07 Mfr # D250QX
COMPARE
250 500 20
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat