Ohaus Defender 7000XW Extreme Washdown Industrial Scales
With rugged construction, comprehensive yet intuitive software, and multiple connectivity options, the Defender 7000 can meet the needs of demanding industrial operations.
  • All stainless steel construction with IP66/NEMA4X protection for indicator, with loadcells protected to IP67
  • Quick display of results using large dual line LCD with high-contrast white backlight
  • Temporary display of 10x readability in weighing mode using quick key press
  • Easy to operate with positive feedback 5-function key and alpha-numeric membrane keypad
  • Weighs in one or more units: kg, g, lb, oz, lb:oz (decimal), tonnes, and user-definable custom unit
  • Multiple weighing modes including counting, percent weighing, dynamic/animal weighing, checkweighing, display hold, and accumulation.
  • 255 item library function
  • Internal power supply with universal line cord
  • Base and indicator connected using quick connect plug
The OHAUS DefenderTM 7000 is a feature-rich line of bench scales, combining a stainless steel D71XW advanced indicator with an all-stainless steel washdown base and a stainless steel column. Ideal for shipping and receiving, production, packaging, and general commercial and industrial weighing applications, the Defender 7000 offers a multifunctional indicator with multiple weighing units, alpha-numeric keypad, and software modes to meet the requirements of industrial and lab applications.
£1,364.00 - £2,531.00GBP / Each
£1,636.80 - £3,037.20GBP (incl VAT)
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus D71XW15HR1 Defender 7000 Rectangle Scale, 30lb x 0.005Lb (15kg x 0.002kg) WZ-11600-87 Mfr # D71XW15HR1
COMPARE
15 30 100 to 240
£1,409.00GBP / Each
£1,690.80GBP (incl VAT)
Ohaus D71XW30HR1 Defender 7000 Rectangle Scale, 60lb x 0.01Lb (30kg x 0.005kg) WZ-11600-88 Mfr # D71XW30HR1
COMPARE
30 60 100 to 240
£1,409.00GBP / Each
£1,690.80GBP (incl VAT)
Ohaus D71XW60HR1 Defender 7000 Rectangle Scale, 150lb x 0.02Lb (60kg x 0.01kg) WZ-11600-89 Mfr # D71XW60HR1
COMPARE
60 150 100 to 240
£1,409.00GBP / Each
£1,690.80GBP (incl VAT)
Ohaus D71XW60HL2 Defender 7000 Rectangle Scale, 150lb x 0.02Lb (60kg x 0.01kg) WZ-11600-90 Mfr # D71XW60HL2
COMPARE
60 150 100 to 240
£1,580.00GBP / Each
£1,896.00GBP (incl VAT)
Ohaus D71XW100HL2 Defender 7000 Rectangle Scale, 250lb x 0.05Lb (100kg x 0.02kg) WZ-11600-91 Mfr # D71XW100HL2
COMPARE
100 250 100 to 240
£1,580.00GBP / Each
£1,896.00GBP (incl VAT)
Ohaus D71XW150HX2 Defender 7000 Rectangle Scale, 300lb x 0.05Lb (150kg x 0.02kg) WZ-11600-92 Mfr # D71XW150HX2
COMPARE
150 300 100 to 240
£1,734.00GBP / Each
£2,080.80GBP (incl VAT)
Ohaus D71XW300HX2 Defender 7000 Rectangle Scale, 600lb x 0.1Lb (300kg x 0.05kg) WZ-11600-93 Mfr # D71XW300HX2
COMPARE
300 600 100 to 240
£1,878.00GBP / Each
£2,253.60GBP (incl VAT)
Ohaus D71XW10WR3 Defender 7000 Square Scale, 25Lb x 0.002Lb (10kg x 0.001kg) WZ-11600-94 Mfr # D71XW10WR3
COMPARE
10 25 100 to 240
£1,364.00GBP / Each
£1,636.80GBP (incl VAT)
Ohaus D71XW25WR3 Defender 7000 Square Scale, 50lb x 0.005Lb (25kg x 0.002kg) WZ-11600-95 Mfr # D71XW25WR3
COMPARE
25 50 100 to 240
£1,387.00GBP / Each
£1,664.40GBP (incl VAT)
Ohaus D71XW50WL4 Defender 7000 Square Scale, 100lb x 0.01Lb (50kg x 0.005kg) WZ-11600-96 Mfr # D71XW50WL4
COMPARE
50 100 100 to 240
£2,150.00GBP / Each
£2,580.00GBP (incl VAT)
Ohaus D71XW100WL4 Defender 7000 Square Scale, 250lb x 0.02Lb (100kg x 0.01kg) WZ-11600-97 Mfr # D71XW100WL4
COMPARE
100 250 100 to 240
£2,150.00GBP / Each
£2,580.00GBP (incl VAT)
Ohaus D71XW250WX4 Defender 7000 Square Scale, 500lb x 0.05Lb (250kg x 0.02kg) WZ-11600-98 Mfr # D71XW250WX4
COMPARE
250 500 100 to 240
£2,531.00GBP / Each
£3,037.20GBP (incl VAT)
Showing 1- 12 of 12